missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta-Galactosidase-1/GLB1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6464-SP
This item is not returnable.
View return policy
Description
beta-Galactosidase-1/GLB1 Monoclonal specifically detects beta-Galactosidase-1/GLB1 in Human samples. It is validated for Western Blot.
Specifications
| beta-Galactosidase-1/GLB1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P16278 | |
| GLB1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human beta-Galactosidase-1/GLB1 Met1-Val677 Accession # P16278 | |
| 25 μg | |
| Primary | |
| Detects human beta-Galactosidase-1/GLB1 in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot | |
| 810226 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Acid beta-galactosidase, beta-galactosidase, betaGalactosidase1, EBP, EC 3.2.1.23, Elastin receptor 1, elastin receptor 1 (67kD), elastin receptor 1, 67kDa, ELNR1, galactosidase, beta 1, GLB1, Lactase, MPS4B | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2720 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction