missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human BAFFR/TNFRSF13C ELISA Kit (Colorimetric)

Product Code. 18644330 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18644330 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18644330 Supplier Novus Biologicals™ Supplier No. NBP269926

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.10 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Biological Fluids
Specificity This kit recognizes Human BAFFR in samples. No significant cross-reactivity or interference between Human BAFFR and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 115650
Gene Symbol TNFRSF13C
Quantity 1 Kit
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym BAFFR;BAFF-R;BROMIX;CD268;CVID4;prolixin;Tumor necrosis factor receptor superfamily member 13 C
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.