missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ATRIP Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF1579-SP
This item is not returnable.
View return policy
Description
ATRIP Polyclonal specifically detects ATRIP in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| ATRIP | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGS1, ATM and Rad3-related-interacting protein, ATR interacting protein, ATR-interacting protein, DKFZp762J2115, FLJ12343, MGC20625, MGC21482, MGC26740, TREX1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 84126 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8WXE1 | |
| ATRIP | |
| E. coli-derived recombinant human ATRIP Glu461-Gly791 Accession # Q8WXE1 | |
| 25 μg | |
| Primary | |
| Detects endogenous human ATRIP in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction