missing translation for 'onlineSavingsMsg'
Learn More

Human ARF3 knockout HEK-293T cell lysate, 1 Kit

Product Code. 32140969 Shop All Abcam Products
This item is not returnable. View return policy
Product Code. 32140969 Supplier Abcam Supplier No. ab2578391Kit
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human ARF3 knockout HEK-293T cell/tissue lysate.

  • Species: Human
  • Tissue: ARF3 knockout HEK-293T
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.