missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Aquaporin 12 (aa 246-290) Control Fragment Recombinant Protein

Product Code. 30204635
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30204635

Brand: Invitrogen™ RP104335

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (42%), Rat (42%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59918 (PA5-59918. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Aquaporin 12 (AQP12) is the most recently identified member of the mammalian AQP family and is specifically expressed in pancreatic acinar cells. In vitro expression studies have revealed that AQP12 is localized at intracellular sites. To determine the physiological roles of AQP12 in the pancreas, we generated knockout mice for this gene (AQP12-KO). No obvious differences were observed under normal conditions between wild-type (WT) and AQP12-KO mice in terms of growth, blood chemistry, pancreatic fluid content, or histology. However, when we induced pancreatitis through the administration of a cholecystokinin-8 (CCK-8) analog, the AQP12-KO mice showed more severe pathological damage to this organ than WT mice.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number A6NM10, Q8IXF9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 375318, 653437
Name Human Aquaporin 12 (aa 246-290) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AQP12; AQP-12; AQP12A; AQP12B; AQP-12 B; AQP x 2; aquaporin 12; aquaporin 12 A; aquaporin 12 B; aquaporin X2; aquaporin-12; aquaporin-12 A; Aquaporin-12 B; INSSA3; insulin synthesis associated 3; RGD1562727
Common Name Aquaporin 12
Gene Symbol AQP12A, AQP12B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FQRNLFYGQKNKYRAPRGKPAPASGDTQTPAKGSSVREPGRSGVE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.