Learn More
R&D Systems™ Human ANO7/NGEP (NP_001001891) VersaClone cDNA
Product Code. 18707399
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18707399 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18707399 Supplier R&D Systems™ Supplier No. RDC2167
  • ONLINE EXCLUSIVE PRICE
    791.00 €
    747.60 € / 10µg
    Save 43.40 €
    5% Off
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
R&D Systems™ Human ANO7/NGEP (NP_001001891) VersaClone cDNA is a cytoplasmic protein.
The Human ANO7/NGEP (NP_001001891) VersaClone cDNA clone contains the complete ORF for ANO7, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The ANO7 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY
Specifications
Gene hANO7
Accession Number NP_001001891
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 2815bp
pH 8.5
Gene ID (Entrez) 50636
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg