missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Annexin A8/ANXA8 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF8105-SP
This item is not returnable.
View return policy
Description
Annexin A8/ANXA8 Polyclonal specifically detects Annexin A8/ANXA8 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Annexin A8/ANXA8 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| annexin A8, Annexin VIII, Annexin-8, Anx8, ANXA8, ANXA8L2, FLJ53095, VAC-beta, Vascular anticoagulant-beta | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 653145 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P13928 | |
| ANXA8 | |
| E. coli-derived recombinant human Annexin A8/ANXA8 Ala2-Pro327 Accession # P13928 | |
| 25 μg | |
| Primary | |
| Detects human Annexin A8/ANXA8 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) ANXA4, rhANXA5, rhANXA6, rhANXA7, rhANXA11, and rhANXA13A is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction