missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiopoietin-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB9231-500
This item is not returnable.
View return policy
Description
Angiopoietin-1 Monoclonal specifically detects Angiopoietin-1 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| Angiopoietin-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, ANGPT1, KIAA0003 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 284 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, ELISA | |
| 171733 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q15389 | |
| ANGPT1 | |
| Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-1 Ser20-Phe498 Accession # Q15389 | |
| 500 μg | |
| Primary | |
| Detects human Angiopoietin-1 in Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) Ang-2, rhAng-4, rmANGPTL3, or rhANGPTL7. | |
| Human | |
| Supernatant |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto