missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Amphiregulin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 16 publications
Brand: R&D Systems AF262-SP
This item is not returnable.
View return policy
Description
Amphiregulin Polyclonal specifically detects Amphiregulin in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry.
Specifications
| Amphiregulin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| amphiregulin, AR, AREG, AREGB, Colorectum cell-derived growth factor, CRDGF, MGC13647, schwannoma-derived growth factor, SDGF | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 374 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.3-1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P15514 | |
| AREG | |
| E. coli-derived recombinant human Amphiregulin Val107-Lys184 Accession # P15514 | |
| 25 μg | |
| Primary | |
| Detects human Amphiregulin in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) Amphiregulin, rmEpiregulin, recombinant human (rh) HB-EGF, and rhTGF-alpha is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction