missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human AMICA/JAML (NP_001091996) VersaClone cDNA

Product Code. 18772799
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18772799 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18772799 Supplier R&D Systems™ Supplier No. RDC1537

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

The Human AMICA/JAML (NP_001091996) VersaClone cDNA clone contains the complete ORF for AMICA/JAML, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The AMICA/JAML insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifications

Gene AMICA/JAML
Accession Number NP_001091996
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.210
Insert Type 1198bp
Gene ID (Entrez) 120425
Gene Alias adhesion molecule, interacts with CXADR antigen 1, AMICA, AMICA1, CREA7-1, Gm638, JAML
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg

For Reasearch Use Only. Not for use in Humans.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.