Learn More
Abnova™ Human ALG6 Partial ORF (NP_037471, 25 a.a. - 114 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00029929-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the ALG6/ALG8 glucosyltransferase family. The encoded protein catalyzes the addition of the first glucose residue to the growing lipid-linked oligosaccharide precursor of N-linked glycosylation. Mutations in this gene are associated with congenital disorders of glycosylation type Ic. [provided by RefSeq]
Sequence: SYSGAGKPPMFGDYEAQRHWQEITFNLPVKQWYFNSSDNNLQYWGLDYPPLTAYHSLLCAYVAKFINPDWIALHTSRGYESQAHKLFMRTSpecifications
NP_037471 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SYSGAGKPPMFGDYEAQRHWQEITFNLPVKQWYFNSSDNNLQYWGLDYPPLTAYHSLLCAYVAKFINPDWIALHTSRGYESQAHKLFMRT | |
RUO | |
ALG6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
29929 | |
ALG6 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALG6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |