Learn More
Abnova™ Human AKAP14 Full-length ORF (NP_848928.1, 1 a.a. - 197 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00158798-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The protein anchors PKA in ciliary axonemes and, in this way, may play a role in regulating ciliary beat frequency. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: MSETQNSTSQKAMDEDNKAASQTMPNTQDKNYEDELTQVALALVEDVINYAVKIVEEERNPLKNIKWMTHGEFTVEKGLKQIDEYFSKCVSKKCWAHGVEFVERKDLIHSFLYIYYVHWSISTADLPVARISAGTYFTMKVSKTKPPDAPIVVSYVGDHQALVHRPGMVRFRENWQKNLTDAKYSFMESFPFLFNRVSpecifications
NP_848928.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
AKAP14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
158798 | |
AKAP14 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AKAP28 | |
AKAP14 | |
Recombinant | |
wheat germ expression system |