missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AHR Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6185
This item is not returnable.
View return policy
Description
AHR Polyclonal specifically detects AHR in Human samples. It is validated for Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated.
Specifications
| AHR | |
| Polyclonal | |
| Unconjugated | |
| P35869 | |
| AHR | |
| E. coli-derived recombinant human AHR Asn704-Leu848 Accession # P35869 | |
| 100 μg | |
| Primary | |
| Detects human AHR in Western blots. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, Immunoassay | |
| LYOPH | |
| Western Blot 1 μg/mL, Simple Western 10 μg/mL, Intracellular Staining by Flow Cytometry 2.5 μg/10^6 cells, Immunocytochemistry 5-15 μg/mL, CyTOF-ready, Knockout Validated | |
| Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 196 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction