missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ADM ELISA Kit

Product Code. 18087404
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 18087404

Brand: Invitrogen™ EEL023

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Adrenomedullin (ADM), a vasodilator produced by most contractile cells, is characterized by persistent hypotensive activity. ADM is involved in the regulation of fluid and electrolyte homeostasis and in the maintenance of cardiovascular functioning. In hypertensive patients, the level of ADM in plasma is upregulated. Natriuresis is a common systemic manifestation of aneurysmal subarachnoid hemorrhage. ADM has strong natriuretic actions. ADM-induced natriuresis is caused by an increase in glomerular filtration rate and a decrease in distal tubular sodium reabsorption. ADM is present both in the periphery and brain, and can exert central effects such as decreasing food ingestion.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35318
Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 133
Gene Symbol ADM
Interassay CV <10%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL, Serum 100 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Adrenomedullin
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.