missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HTR4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15961825
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15961825 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15961825 Supplier Invitrogen™ Supplier No. PA575300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene is a member of the family of serotonin receptors, which are G protein coupled receptors that stimulate cAMP production in response to serotonin (5-hydroxytryptamine). The gene product is a glycosylated transmembrane protein that functions in both the peripheral and central nervous system to modulate the release of various neurotransmitters. Multiple transcript variants encoding proteins with distinct C-terminal sequences have been described, but the full-length nature of some transcript variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene HTR4
Gene Accession No. P97288, Q13639, Q62758
Gene Alias 5 hydroxytryptamine ( serotonin) receptor 4; 5 hydroxytryptamine (serotonin) receptor 4; 5-HT>4L>; 5HT4; 5-HT4; 5-HT-4; 5HT4 receptor; 5-HT4 receptor; 5-HT4 receptor (two splicing variants L and S); 5-HT4R; 5HTR4; 5-hydroxytryptamine (serotonin) receptor 4, G protein-coupled; 5-hydroxytryptamine receptor 4; cardiac 5-HT4 receptor; Htr4; Serotonin receptor 4
Gene Symbols HTR4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 14-66 of Human HTR4.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15562, 25324, 3360
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.