missing translation for 'onlineSavingsMsg'
Learn More

HT-2157, MedChemExpress

Product Code. 16493128
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493128 10 mg 10mg
16449686 50 mg 50mg
16435867 5 mg 5mg
16434426 100 mg 100mg
16420689 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493128 Supplier Medchem Express Supplier No. HY100717

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HT-2157 (SNAP 37889) is a selective, high-affinity, competitive antagonists of galanin-3 receptor (Gal3).

HT-2157 (SNAP 37889) is a selective, high-affinity, competitive antagonists of galanin-3 receptor (Gal3).

Specifications

Chemical Name or Material HT-2157
Physical Form Solid
CAS 303149-14-6
Color Orange
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C21H13F3N2O
For Use With (Application) Neuroscience-Neuromodulation
Synonym SNAP 37889
Solubility Information DMSO : ≥ 29 mg/mL (79.16 mM)
SMILES O=C1N(C2=CC=CC=C2)C3=C(C=CC=C3)/C1=N/C4=CC=CC(C(F)(F)F)=C4
Molecular Weight (g/mol) 366.34
Formula Weight 366.34
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.