missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33501
This item is not returnable.
View return policy
Description
HSPA2 Polyclonal specifically detects HSPA2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| HSPA2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| HSPA2 | |
| Recombinant protein encompassing a sequence within the center region of human HSPA2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.34mg/mL | |
| Western Blot 1:500-1:3000 | |
| Heat shock 70 kDa protein 2, heat shock 70kD protein 2, heat shock 70kDa protein 2, heat shock-related 70 kDa protein 2, HSP70-2, HSP70-3 | |
| Rabbit | |
| 70 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 3306 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction