Learn More
HSP27 Rat anti-Human, Clone: 836370, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB23142-SP
Description
HSP27 Monoclonal specifically detects HSP27 in Human samples. It is validated for Western Blot.Specifications
| HSP27 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HSPB1, SRP27, Stress-responsive protein 27 | |
| Phosphopeptide containing human HSP27 S78/S82 sites Accession # P04792 | |
| 25 μg | |
| Primary | |
| Detects human HSP27 when dually phosphorylated at S78/S82. | |
| Human | |
| Purified |
| Western Blot | |
| 836370 | |
| Western Blot 0.25 ug/mL | |
| P04792 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3315 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only (RUO)