missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP27 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
200.00 €
Specifications
| Antigen | HSP27 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000 - 1:2000, Immunohistochemistry 4 μg/mL, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
HSP27 Polyclonal specifically detects HSP27 in Human, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| HSP27 | |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry 4 μg/mL, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience | |
| Constituents: PBS, 1mg/ml BSA with 0.05% Sodium Azide | |
| 3315 | |
| Synthetic peptide corresponding to residues L(10) L R G P S W D P F R C(21) of human Heat Shock Protein 27. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
| HSPB1 | |
| IgG | |
| Affinity Purified | |
| Hsp27 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title