missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP27 Antibody (6H11), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25149IR
This item is not returnable.
View return policy
Description
HSP27 Monoclonal specifically detects HSP27 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
| HSP27 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HSPB1 | |
| Recombinant full length human HSP27 expressed in and purified from E. coli. [UniProt# P04792] | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific | |
| 3315 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 6H11 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse (Negative), Rat (Negative) | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction