missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSFY1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 18015591
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18015591 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18015591 Supplier Invitrogen™ Supplier No. PA5145084

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: STRSPLCEHT FPGDSDLRSM IEEHAFQVLS QGSLLESPSY TVCVSEPDKD DDFLSLNFPR KLWKIVESDQ FKS Antigen sequence identity: Mouse 56%; Rat 54%.

This gene encodes a member of the heat shock factor (HSF) family of transcriptional activators for heat shock proteins. This gene is a candidate gene for azoospermia, since it localizes to a region of chromosome Y that is sometimes deleted in infertile males. The genome has two identical copies of this gene within a palindromic region; this record represents the more centromeric copy. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSFY1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HSFY1
Gene Accession No. Q96LI6
Gene Alias Heat shock transcription factor 2-like protein; heat shock transcription factor, Y-linked; heat shock transcription factor, Y-linked 1; HSF2L; HSF2-like; HSFY; HSFY1; HSFY2; Y-linked 1
Gene Symbols HSFY1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 23-95. Recombinant protein control fragment (Product #RP-110156).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 86614
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.