missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The HSD3B2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human HSD3B2. This antibody recognizes human, mouse, rat antigen. The HSD3B2 antibody has been validated for the following applications: WB, IHC, FC (Intra), ELISA analysis.
Specifications
Specifications
| Antigen | HSD3B2 |
| Applications | Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Gene Symbols | HSDB; HSDB3; SDR11E2 |
| Host Species | Rabbit |
| Immunogen | Recombinant Protein |
| Purification Method | Antigen affinity purification |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?