missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPRT Antibody (OTI2F7), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70934PE
This item is not returnable.
View return policy
Description
HPRT Monoclonal antibody specifically detects HPRT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| HPRT | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry | |
| OTI2F7 | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| EC 2.4.2.8, HGPRTHPRTHGPRTase, hypoxanthine phosphoribosyltransferase 1, hypoxanthine-guanine phosphoribosyltransferase | |
| Full length human recombinant protein of human HPRT1(NP_000185) produced in HEK293T cell. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 3251 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction