Learn More
Thermo Scientific™ HPLC Qualification Kits for UltiMate™ 3000 Systems
Product Code. 17834494 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
OP/PQ Standards Refill Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
17834494 OP/PQ Standards Refill Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17834494 Supplier Thermo Scientific™ Supplier No. 3323.0010
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
HPLC Qualification Kits are for system validation of UltiMate 3000 LC and UHPLC systems.
Choose Thermo Scientific™ Dionex™ UltiMate™ HPLC Qualification Kits for system validation of UltiMate 3000 LC and UHPLC systems.
The modules of UltiMate 3000 RSLCnano system can be arranged in different configurations for various applications.
  • Components include fittings, capillaries and columns, but vary by application
Specifications
Description OP/PQ Standards Refill Kit
For Use With (Equipment) Dionex UltiMate 3000 Series
Product Line UltiMate