Learn More
Justrite™ HPLC Poly Manifold Fittings for Quick-Disconnect Disposal Cans
Specifications
Specifications
| Certifications/Compliance | US OSHA, NFPA Code 30 |
| Diameter (Metric) Outer | 0.2 cm |
| Diameter (English) Outer | 0.625 in. |
| Includes | 6 connectors |
| For Use With (Equipment) | HPLC poly manifold, tube size 0.062 in. O.D., 0.031 in. I.D. |
| Packaging | Each |
| Material | Resin |
| Product Type | Quick-disconnect Disposal Can |