Learn More
HPGDS inhibitor 2, MedChemExpress
Product Code. 16487977
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16487977
Brand: Medchem Express HY126134
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress HPGDS inhibitor 2 is a highly potent and selective hematopoietic prostaglandin D synthase (H-PGDS) inhibitor with an IC50 of 9.9 nM.
HPGDS inhibitor 2 is a highly potent and selective hematopoietic prostaglandin D synthase (H-PGDS) inhibitor with an IC50 of 9.9 nM.
Specifications
Chemical Name or Material HPGDS inhibitor 2
CAS 2101626-26-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H24F2N2O3
Quantity 5 mg
Solubility Information DMSO : ≥ 150 mg/mL (396.40 mM)
SMILES FC(F)OC1=CC=C2C(N=CC(C(N[C@@H]3CC[C@@H](C(C)(C)O)CC3)=O)=C2)=C1
Molecular Weight (g/mol) 378.41
Formula Weight 378.41
Percent Purity 99.72%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation