missing translation for 'onlineSavingsMsg'
Learn More

HPGDS inhibitor 1, MedChemExpress

Product Code. 16446687
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16446687

Brand: Medchem Express HY10439

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HPGDS inhibitor 1 is a potent, selective and orally active Hematopoietic Prostaglandin D Synthase (HPGDS) inhibitor with an IC50s of 0.6 nM and 32 nM in enzyme and cellular assays, respectively. HPGDS inhibitor 1 does not inhibit human L-PGDS, mPGES, COX-1, COX-2, or 5-LOX.

HPGDS inhibitor 1 is a potent, selective and orally active Hematopoietic Prostaglandin D Synthase (HPGDS) inhibitor with an IC50s of 0.6 nM and 32 nM in enzyme and cellular assays, respectively. HPGDS inhibitor 1 does not inhibit human L-PGDS, mPGES, COX-1, COX-2, or 5-LOX.

Specifications

Chemical Name or Material HPGDS inhibitor 1
CAS 1033836-12-2
Color White
Purity Grade Notes Research
Molecular Formula C19H19F4N3O
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (131.11 mM; Need ultrasonic)
SMILES O=C(NC1CCN(CC(F)(F)F)CC1)C2=CC=C(C3=CC=CC(F)=C3)N=C2
Molecular Weight (g/mol) 381.37
Formula Weight 381.37
Percent Purity 99.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.