Learn More
Agilent Technologies HP-PLOT-Q 30m, 0.32mm, 20um
Brand:  Agilent Technologies 19091P-QO4
Encompass_Preferred
HP-PLOT-Q 30m, 0.32mm, 20um
Product Code. 10172683
  • 1070.85 € / Each
Only null left
Estimated Shipment: 16-01-2026