Learn More
Agilent Technologies HP-1 15M 0.25mm 1.0U
Brand:  Agilent Technologies 19091Z-231
Encompass_Preferred
HP-1 15M 0.25mm 1.0U
Product Code. 10000633
  • 540.87 € / Each
Only null left
Estimated Shipment: 13-01-2026