missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HOXA4 Polyclonal Antibody

Product Code. 15994955
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15994955 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15994955 Supplier Invitrogen™ Supplier No. PA579389

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: SW620 Cell, SW620 Cell, PC-12 Cell Nuclear. IHC: Human Mammary Cancer Tissue. ICC/IF: HELA Cell.

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene HOXA4
Gene Accession No. P06798, P09635, Q00056
Gene Alias AV206827; Dfd-like protein; homeo box A4; homeobox A4; homeobox protein Hox-1.4; Homeobox protein Hox-1D; homeobox protein Hox-A4; homeobox protein Hox-A4-like; homeobox protein MH-3; HOX1; Hox-1.4; Hox-1.4-like protein; Hox-14; HOX1D; Hoxa4; Hoxa-4; LOC100912525
Gene Symbols HOXA4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human HOXA4 (9-26aa NSNYIEPKFPPFEEYAQH).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100912525, 15401, 3201
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.