Learn More
Gestigkeit Harry Hotplate with Performance Control and Thermostatic Controller
Click to view available options
Dimensions (L x W x H):
358 x 358 x 170 mm
592 x 442 x 170 mm
Temperature (Metric):
130°C to 370°C
30°C to 110°C
50°C to 300°C
Weight (Metric):
13 kg
26 kg
Voltage:
230 V
400 V
Unit Size:
Each
Description
- Electrical hotplates with variable temperature control, with or without a thermostatic controller. Robust construction for continuous perfomance. aluminum alloy hotplate.
- Because of the asymmetrical, long-term heating system (hotplate corners and edges are subjected to more heat) an even temperature is guaranteed over the entire heating surface.
- Asbest-free insulation. The housing is constructed from high-grade, stainless steel No. 1.4301, whereby the central section has been additionally sprayed in orange.
- Four adjustable feet ensure extra stability. Connecting cable approx. 1,7 m. long, 230 Volt/ 3200 Watt with Schuko plug.
Specifications
Specifications
| Body Material | Stainless Steel |
| Top Plate Material | Anodized Aluminum |
| Temperature (Metric) | 130°C to 370°C |
| Weight (Metric) | 26 kg |
| Wattage | 4000 W |
| Includes | Performance Control and Thermostatic Controller |
| Voltage | 230 V |
| Type | Hotplate |
| Dimensions (L x W x H) | 592 x 442 x 170 mm |