Learn More
Gestigkeit Harry™ Hotplate, 50°C to 500°C, Ceramic
Product Code. 10222491
Click to view available options
Top Plate Material:
Ceramic
Unit Size:
Each
Product Code. 10222491
Brand: Gestigkeit Harry™ 33A
Only null left
Add to basket
Add to basket
Shape: Square, Size: 16.9 x 16.9 in.
Shape: Square; Size: 16.9 x 16.9 in
Shape: Square; Size: 16.9 x 16.9 in.
Specifications
Top Plate Material Ceramic
Size 16.9 x 16.9 in.
Temperature (Metric) 50°C to 500°C
Depth (Metric) 440 mm
Weight (English) 19.8 lb.
Height (Metric) 100 mm
Weight (Metric) 9 kg
Width (Metric) 440 mm
Wattage 4400 W
Electrical Requirements 230 V, 50/60 Hz, 3000 w
Frequency 50/60 Hz
Shape Square
Voltage 400 V
Type Hot Plate