Learn More
Buerkle™ Polypropylene Hose Nozzle for Buerkle™ Stopcock
Shop All Bürkle Products
Click to view available options
Diameter (Metric):
8 mm
12 mm
Unit Size:
Each
Description
- Media-contacting parts constructed of Polypropylene, Polyethylene, and PTFE
- Available in a variety of sizes
Specifications
Specifications
| Type | Hose Nozzle |
| Diameter (Metric) | 12 mm |
| Description | Blue |
| Material | Polypropylene |
| For Use With (Equipment) | StopCock Discharge Cock |