Learn More
Medicom™ Hopen™ Collared Hygiene Gown
Medicom™ Hopen™ Polypropylene Coveralls available in collared, or hooded styles
Brand:  Medicom™ 40.408/03
 View more versions of this product
Product Code. 16584061
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • Pleasant, and light PP SMS fabric is breathable and solid
  • SMS fabric makes it possible to better limit the particulate release of fibers
  • Category 1 Personal Protective Equipment
  • Suitable for protected environments
  • Individually packaged for better hygiene and controlled distribution
  • Elastic thumb holes contain latex, a natural material that contains allergenic proteins
  • Dark blue gown has anti-static properties giving it the ability to dissipate electrostatic charges
    • Not intended for use in a flammable, oxygen-enriched atmosphere, or an ATEX area
Specifications
Specifications
Large
Polypropylene, SMS
Zipper
Non-sterile
No
Manufactured under an ISO 9001 Certified Quality System
Personal Protective Equipment: Category 1
50 g/m2
Unisex
White
Yes
1
Sewn
EN 1149-5
Individual, Bag