Learn More
Honeywell KCL™ Honeywell KCL™ Vitoject™ 890 Gloves
Click to view available options
Size:
9
10
11
Unit Size:
1 pair
Description
- AQL < 0.65
- Loose fit allows heat-protecting gloves to be worn underneath
- Can be re-used after contamination with certain chemicals, after special cleaning process (please consult KCL Laboratories)
- Can be combined with chemical protective clothing
- Excellent low degradation
- Good resistance against polycyclic aromatic hydrocarbons (petrochemicals
Specifications
Specifications
| Certifications/Compliance | IFA 1301115 |
| Color | Black |
| Latex Free | Yes |
| Material | Viton |
| Powdered | Powder Free |
| Product Type | Gloves |
| Size | 11 |
| Thickness (Metric) | 0.7 cm |
| Length (Metric) | 340-360 cm |