Learn More
Edmund Bühler GmbH™ Homogenizer Knife
Product Code. 10595621
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
150 mL
Unit Size:
Each
Product Code. 10595621
Brand: Edmund Bühler GmbH™ 0002 987
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from stainless steel. Edmund Bühler GmbH™ Homogenizer Knife is for use with a variety of sample vessels.
  • Provides mechanical crushing of samples used with Homogenizer HO 4/A
  • Stainless steel construction
  • Spindle, knife and sample vessel must be matched in size.
Specifications
Product Type Homogenizer Knife
Capacity (Metric) 150 mL
Blade Material Stainless Steel