missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Hoechst 33258

Product Code. p-200104459
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16432717

Brand: MedChemExpress HY15558

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hoechst 33258 is a fluorescent dye that emits blue fluorescence when bound to dsDNA.

Specifications

Content And Storage -20°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Quantity 50 mg
Shipping Condition Room temperature
Solubility DMSO : 41.67 mg/mL (98.16 mM; ultrasonic and warming and heat to 60°C)
Molecular Weight (g/mol) 424.5 g/mol
CAS 23491-44-3
Synonym bisBenzimide H 33258 ∣ H 33258
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.