missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HNF4G Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590904
This item is not returnable.
View return policy
Description
Immunogen sequence: ASNDGSHLHH PMHPHLSQDP LTGQTILLGP MSTLVHADQI STPETPLPSP PQGSGQEQYK IAANQASVIS HQHLSKQKQL; Positive Samples: HT-29, Mouse pancreas, Rat pancreas, Rat kidney; Cellular Location: Nucleus.
HNF4G is a transcription factor. It has a lower transcription activation potential than HNF4-alpha.
Specifications
| HNF4G | |
| Polyclonal | |
| Unconjugated | |
| HNF4G | |
| hepatocyte nuclear factor 4 gamma; hepatocyte nuclear factor 4, gamma; hepatocyte nuclear factor 4-gamma; HNF4 gamma; HNF-4 gamma; HNF4-beta; Hnf4g; Hnf4gamma; HNF-4-gamma; NR2A2; NR2A3; Nuclear receptor subfamily 2 group A member 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 30942, 3174, 365744 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.61 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q14541, Q9WUU6 | |
| HNF4G | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 366-445 of human HNF4G (NP_0041244?). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction