Learn More
Invitrogen™ HMGB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578006
Description
Positive Control: 293T, A431, HeLa, HepG2, human HMGB1-transfected 293T, human HMGB2-transfected 293T Predicted Reactivity: Mouse (100%), Rat (100%), Pig (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HMGB2 encodes a member of the non-histone chromosomal high mobility group protein family. The proteins of this family are chromatin-associated and ubiquitously distributed in the nucleus of higher eukaryotic cells. In vitro studies have demonstrated that this protein is able to efficiently bend DNA and form DNA circles. These studies suggest a role in facilitating cooperative interactions between cis-acting proteins by promoting DNA flexibility. This protein was also reported to be involved in the final ligation step in DNA end-joining processes of DNA double-strand breaks repair and V(D)J recombination.
Specifications
| HMGB2 | |
| Polyclonal | |
| Unconjugated | |
| Hmgb2 | |
| C80539; high mobility group box 2; high mobility group protein 2; High mobility group protein B2; high-mobility group (nonhistone chromosomal) protein 2; high-mobility group box 2; HMG 2; HMG B2; Hmg2; HMG-2; HMGB2; HMG-B2; HMGB-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3148 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.7 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P26583 | |
| Hmgb2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human HMGB2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.