Learn More
HLA G Antibody (MEM-G/9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NB500-314
Product Code. 18719243
  • 460.00 € / 0.10mg
Please to purchase this item. Need a web account? Register with us today!
Description
Description
HLA G Monoclonal specifically detects HLA G in Human, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monoclonal
Store at 4C. Do not freeze.