missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR/Monocyte Mouse anti-Human, PE,PerCP-Cy™,¢5.5, Clone: L243,MφP9 (also known as MφP-9), BD Quantibrite™
Mouse Monoclonal Antibody
Brand: BD Biosciences 340827
This item is not returnable.
View return policy
Description
HLA-DR/Monocyte Monoclonal antibody specifically detects HLA-DR/Monocyte in Human samples. It is validated for Flow Cytometry,Fluorescent Quenching.
Specifications
| HLA-DR/Monocyte | |
| Monoclonal | |
| PE/PerCP-Cyanine5.5 | |
| 50 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2a κ, IgG2b κ |
| Flow Cytometry | |
| L243, MφP9 (also known as MφP-9) | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction