missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DQW1 Antibody (Genox 3.53), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2702AF405
This item is not returnable.
View return policy
Description
HLA DQW1 Monoclonal specifically detects HLA DQW1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| HLA DQW1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| CELIAC1, DC1, HLA class II histocompatibility antigen, DQ beta 1 chain, HLA-DQB, IDDM1, LB-E12, MB1, MHC class II antigen DQB1, MHC class II antigen HLA-DQ-beta-1, MHC class II DQ beta chain, MHC class II HLA-DQ beta glycoprotein, MT1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human Leukocyte Antigen DQW1 (DC1 determinant of human HLA-DR antigens) | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Genox 3.53 | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| HLA-DQB1 | |
| Bristol 8 glycoprotein from the Bristol 8 B lymphoblastoid cell line. | |
| 0.1 mL | |
| Asthma, Diabetes Research, Immunology | |
| 3119 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction