Learn More
Epredia™ HistoScreen™ Cassettes in Tube Packs, aqua
Product Code. 12568966 Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Each
Pack of 750
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
12568966 Orange Pack of 750
10168528 Aqua Each
10128578 Blue Each
12588966 Green Pack of 750
10464530 Gray Each
10066878 Lilac Each
10683059 Pink Each
10128008 Tan Each
10466450 White Each
12578966 Yellow Pack of 750
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 12568966 Supplier Epredia™ Supplier No. C750OR
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Handle small biopsies and fragile specimens with Epredia™ HistoScreen™ Cassettes in Tube Packs.
  • Saves time, offers the greatest specimen security, and provides ease of use
  • Provides an inner chamber lined with a “see-through” mesh for easy identification of specimens
  • Eliminates the need for messy sponges or lens paper
  • Ensures your tissues are safely processed with cassettes which will not pop open during processing
TRUSTED_SUSTAINABILITY
Specifications
Color Orange