Description
- Saves time, offers the greatest specimen security, and provides ease of use
- Provides an inner chamber lined with a “see-through” mesh for easy identification of specimens
- Eliminates the need for messy sponges or lens paper
- Ensures your tissues are safely processed with cassettes which will not pop open during processing
Specifications
Specifications
| Pore Style | Mesh Pore Insert |
| Color | Aqua |
| Product Type | HistoScreen™ Cassette |