Learn More
Histone H4, p Ser1 Antibody (RM194), Novus Biologicals™
Product Code. 18660487 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18660487 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18660487 Supplier Novus Biologicals Supplier No. NBP261553
Please to purchase this item. Need a web account? Register with us today!
Rabbit Monoclonal Antibody
Histone H4 Monoclonal antibody specifically detects Histone H4 in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay
TRUSTED_SUSTAINABILITY
Specifications
Antigen Histone H4
Applications Western Blot, ELISA, Immunofluorescence, Multiplex
Classification Monoclonal
Clone RM194
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 μg/mL-2 μg/mL, ELISA 0.2 μg/mL - 1 μg/mL, Immunocytochemistry/ Immunofluorescence 1 μg/mL - 2 μg/mL, Multiplex Immunoassay 0.1 μg/mL - 1 μg/mL
Formulation 50% Glycerol/PBS, 1% BSA
Gene Alias H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
Host Species Rabbit
Immunogen A phospho-peptide corresponding to Histone H4 [p Ser1]
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 121504
Target Species Human, Vertebrates
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG