missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H4 Mouse anti-Human, Clone: KM-2, Novus Biologicals™
Mouse Monoclonal Antibody
244.00 € - 658.00 €
Specifications
| Antigen | Histone H4 |
|---|---|
| Clone | KM-2 |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18316114
|
Novus Biologicals
NBP3-12018-0.025MG |
0.025 mg |
244.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18377281
|
Novus Biologicals
NBP3-12018-0.2MG |
0.2 mg |
658.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H4 Monoclonal antibody specifically detects Histone H4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
| Histone H4 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 | |
| KM-2 antibody was derived from an MRL/lpr lupus mouse. | |
| Protein A purified |
| KM-2 | |
| Monoclonal | |
| Mouse | |
| Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Phospho Specific | |
| PBS | |
| 121504 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title