missing translation for 'onlineSavingsMsg'
Learn More

Histone H3, p Thr45 Antibody, Novus Biologicals™

Product Code. 18258152 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18258152 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18258152 Supplier Novus Biologicals Supplier No. NBP262795

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Histone H3 Polyclonal specifically detects Histone H3 in Human samples. It is validated for ELISA, Dot Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HistoneH3
Applications Dot Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:5000 - 1:10000, Dot Blot 1:20000
Formulation PBS with 0.05% Sodium Azide and 0.05% ProClin 300
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3T45p, HIST2H3C, histone H3.3, MGC87782, MGC87783
Gene Symbols H3C14
Host Species Rabbit
Immunogen This Histone H3 [p Thr45] antibody was raised against histone H3 containing the phosphorylated threonine 45 (H3T45p)
Molecular Weight of Antigen 15 kDa
Purification Method Affinity Purified
Quantity 50 μg
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.