missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, Methyl Lys4 Antibody (C.15200150), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59284
This item is not returnable.
View return policy
Description
Histone H3 Monoclonal specifically detects Histone H3 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Histone H3 (Methyl Lys4) | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| H3C14 | |
| The exact sequence of the immunogen to this Histone H3 [Methyl Lys4] antibody (C.15200150) is proprietary. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, ChIP Assay | |
| C.15200150 | |
| Western Blot 1:1000, ELISA 1:5000, Immunocytochemistry/Immunofluorescence 1:500, Chromatin Immunoprecipitation (ChIP) 1-5 ug | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K4Me, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| Mouse | |
| 15 kDa | |
| 50 μg | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction