missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2a Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
236.00 € - 422.00 €
Specifications
| Antigen | Histone H2a |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 32849530), ELISA 1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18074085
|
Novus Biologicals
NB100-56346SS |
0.025 mg |
236.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18741334
|
Novus Biologicals
NB100-56346 |
0.1 mg |
422.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H2a Polyclonal specifically detects Histone H2a in Human, Mouse, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Histone H2a | |
| Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 32849530), ELISA 1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Sheep | |
| P04908 | |
| 8338 | |
| This antibody was generated by immunizing rabbits with a mixture of synthetic peptides containing amino acid residues 1-15 (MSGRGKQGGKARAKA) and 81-96 (PRHLQLAIRNDEELNK) of human Histone H2A. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, Epigenetics, Histones and Modified Histones | |
| PBS with 0.05% Sodium Azide | |
| H2A histone family, member Q, H2A/q, H2AFQH2A, H2A-GL101, histone 2, H2ac, histone cluster 2, H2ac, histone H2A type 2-C, Histone H2A/q, Histone H2A-GL101, histone IIa, MGC74460 | |
| HIST2H2AC | |
| IgG | |
| Protein G purified | |
| A 14 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title