missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H1 Antibody (HH1/957), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47754F
This item is not returnable.
View return policy
Description
Histone H1 Monoclonal antibody specifically detects Histone H1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifications
| Histone H1 | |
| Monoclonal | |
| FITC | |
| H1 histone family, member 1, H1.1, H1A, H1F1HIST1, histone 1, H1a, histone cluster 1, H1a, histone H1.1, MGC126642, MGC138345 | |
| Recombinant full-length human H1F0 Histone H1.0 protein | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, Epigenetics, Nuclear Envelope Markers | |
| 3024 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| HH1/957 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction